Order Entry
Sweden
ContactUsLinkComponent
Iron(III) oxide
Iron(III) oxide
Catalog # SWED6792
Supplier:  SWED HANDLING
CAS Number:  
Iron(III) oxide
Catalog # SWED6792
Supplier:  SWED HANDLING
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :159.69 g/mol
Merck Index:13,04058
Melting Pt:1538 °C
Density:5.24 g/cm³ (20 °C)
EINECS:215-168-2
 :GHS07
CAS Number:1309-37-1
Formula:Fe₂O₃
MDL Number:MFCD00011008