Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iron (III) oxide
Iron(III) oxide, powder, NanoArc® 20-40nm APS, S.A. 30-60m2/g for cosmetic
Iron(III) oxide, powder, NanoArc® 20-40nm APS, S.A. 30-60m2/g for cosmetic
:
Thermo Fisher Scientific
Iron(III) oxide, powder, NanoArc® 20-40nm APS, S.A. 30-60m2/g for cosmetic
:
Thermo Fisher Scientific
:
1309-37-1
:
:
Fe₂O₃
:
MFCD00011008
:
159.69 g/mol
:
13,04058
:
1565 °C
:
5.24
:
215-168-2
:
GHS07
:
Ambient
msds_ALFA044895
Thermo Scientific Acroseal 25 Years