

Manganese(IV) oxide ≥90% (RT), Technical Grade, activated, Fluka™
:1313-13-9
:
:MnO₂
:MFCD00003463
:86.94 g/mol
:14,05730
: Dec.
:5.21 g/cm³ (21 °C)
:215-202-6
:GHS08
:GHS07
:Ambient
- Appearance:complying
- Identity:complying
- Assay:Min. 90.0 %
- Loss on drying (105°C):Max. 1.5 %