Vanadium(III) chloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7718-98-1
:
Formula:Cl₃V
MDL Number:MFCD00011454
:157.3 g/mol
Melting Pt:250 °C (dec.)
Density:3 g/cm³
Storage Temperature:Ambient
EINECS:231-744-6
UN:2475
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Dark blue to violet to black Powder
- Titration Argentometric:≥96.0 %
- Insoluble matter:≤0.5 % (in water)