Order Entry
Sweden
ContactUsLinkComponent
Zinc chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Zinc chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Zinc chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    4320 g/l (25 °C)
  • Melting Point:
    283 °C
  • Molar Mass:
    136.30 g/mol
  • Bulk Density:
    1400 - 1800 kg/m3
  • Boiling Point:
    732 °C (1013 hPa)
  • Vapor Pressure:
    1.33 hPa (428 °C)
  • Density:
    2.91 g/cm3 (25 °C)
  • pH:
    5 (100 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information