Copper(II) sulphate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
ADR:9,III
:M-Clarity™ quality level = MQ300
UN:3077
:GHS07
:GHS09
Specification Test Results
- Solubility:203 g/l (20 °C)
- Molar Mass:159.61 g/mol
- Bulk Density:800 kg/m3
- Density:3.60 g/cm3 (20 °C)
- pH:3.5 - 4.5 (50 g/l, H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_102791
SDS
ProductPage_102791 ProductPage_102791