Order Entry
Sweden
ContactUsLinkComponent
Copper(II) oxide, Supelco®
Copper(II) oxide, Supelco®
Supplier:  Merck
Certificates
Copper(II) oxide, Supelco®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    1326 °C (decomposition)
  • Molar Mass:
    79.55 g/mol
  • Bulk Density:
    500 kg/m3
  • Density:
    6.48 g/cm3 (25 °C)
  • pH:
    7 (50 g/l, H2O, 20 °C) (slurry)

Specification Test Results

Related Information