Cerium(IV) sulphate
Catalog # GFSC24
Supplier: GFS CHEMICALS
CAS Number:
Specifications
- Förp.:500 g
- Seq:3645
Specifications
About this item
:
:332.24 g/mol
Merck Index:12,02036
Melting Pt:195 °C
Storage Temperature:Omgivande
EINECS:237-029-5
UN:3260
ADR:8,III
:GHS07
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852