
Amidosulphonic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:5329-14-6
:
Formula:H₃NO₃S
MDL Number:MFCD00011603
:97.09 g/mol
Merck Index:13,09006
Boiling Pt:247 °C (1013 hPa)
Melting Pt:205 °C
Density:2.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-218-8
UN:2967
ADR:8,III
:GHS07
Specification Test Results
- Assay:Min. 99.5 %
- Heavy metals (as Pb):Max. 5 ppm
- Ignition residue:Max. 0.02 %
- Cl (Chloride):Max. 10 ppm
- NO3 (Nitrate):Max. 20 ppm
- SO4 (Sulphate):Max. 0.1 %
- Fe (Iron):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC20672.187 spec_VWRC20672.187 spec_VWRC20672.234 spec_VWRC20672.234 spec_VWRC20672.234
Product specification
More