Order Entry
Sweden
ContactUsLinkComponent
Ammonium sulphate ≥99.0% Reag. Ph. Eur.
Ammonium sulphate ≥99.0% Reag. Ph. Eur.
Catalog # 85509.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium sulphate ≥99.0% Reag. Ph. Eur.
Catalog # 85509.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Appearance:
    White crystals
  • pH (5 %):
    4.5 - 6.0
  • Sulphated ash:
    Max. 0.1 %

Specification Test Results

About this item

 :
 :132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391