Ammonium sulphate ≥99.95% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-20-2
:
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391
:132.14 g/mol
Merck Index:14,555
Melting Pt:∼ 280 °C (dec.)
Density:1.769
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46