Order Entry
Sweden
ContactUsLinkComponent
Ammonium chloride 99.8-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Irritant
Ammonium chloride 99.8-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Ammonium chloride 99.8-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calc. on dried substance):
    99.8 - 100.5 %
  • Acidity or alkalinity:
    Passes test Ph.Eur.
  • Appearance of solution:
    Passes test Ph.Eur.
  • Bromides and iodides:
    Passes test Ph.Eur.
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • pH (25°C; 5 %):
    4.5 - 5.5
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (105°C; 2 h):
    Max. 1 %
  • NO3 (Nitrate):
    Max. 5 ppm
  • PO4 (Phosphate):
    Max. 2 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Ca (Calcium):
    Max. 5 ppm
  • Cu (Copper):
    Max. 2 ppm
  • Fe (Iron):
    Max. 2 ppm
  • K (Potassium):
    Max. 50 ppm
  • Mg (Magnesium):
    Max. 5 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Ni (Nickel):
    Max. 1 ppm
  • Pb (Lead):
    Max. 1 ppm
  • Zn (Zinc):
    Max. 2 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results