Benzyltriethylammonium chloride 98%
:56-37-1
:
:C₁₃H₂₂ClN
:MFCD00011824
:227.78 g/mol
:185 °C (dec.)
:>275 °C
:Ambient
:200-270-1
:4.1,III
:GHS07
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:97.5 to 102.5 %
- Water:≤2.0 % (K.F.)