

Triisopropylsilane 98%
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:6485-79-6
:
Formel:C₉H₂₂Si
MDL-nummer:MFCD00009657
:158.36 g/mol
Kokande Pt:84…86 °C (35 mmHg)
Densitet:0.773 g/cm³
Flash Pt:37 °C
Förvaringstemperatur:Ambient
FN:1993
ADR:3,III
:GHS07
:GHS02
Specifikationstestresultat
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4360 (20°C, 589 nm)
- GC:≥97.5 %