


Bromotrimethylsilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2857-97-8
:
Formula:C₃H₉BrSi
MDL Number:MFCD00000048
:153.09 g/mol
Boiling Pt:79 °C (1013 hPa)
Melting Pt:–43 °C
Density:1.1895 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:220-672-0
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
UN:2920
:GHS02
:GHS05
:GHS07
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:-43 °C
- Molar Mass:153.09 g/mol
- Boiling Point:79 °C
- Flash Point:32 °C
- Refractive Index:1.4211 (25 °C, 589 nm)
- Density:1.17 g/cm3 (20 °C)