Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
Vinyltrimethoxysilane for synthesis, Sigma-Aldrich®
  8.41268.0100
 :  Merck
 :  
 
 
 
Vinyltrimethoxysilane for synthesis, Sigma-Aldrich®
  8.41268.0100
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Molar Mass:
    148.23 g/mol
  • Boiling Point:
    122 - 124 °C
  • Vapor Pressure:
    88 hPa (55 °C)
  • Flash Point:
    22 °C
  • Explosion Limit:
    1.1 %(V)
  • Density:
    0.97 g/cm3 (20 °C)
  • Ignition Point:
    235 °C

 

 

 :
 :148.23 g/mol
 :123 °C (1013 hPa)
 :0 °C
 :0.97 g/cm³ (20 °C)
 :Ambient
 :220-449-8
 :1993
 :3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS08
 :GHS07
 :2768-02-7
 :C₅H₁₂O₃Si
 :MFCD00008605