
Germanium(IV) oxide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-53-8
:
Formula:GeO₂
MDL Number:MFCD00011030
:104.61 g/mol
Merck Index:14,4409
Melting Pt:1086 °C
Density:6.239
EINECS:215-180-8
:GHS07
Storage Temperature:Ambient