Order Entry
Sweden
ContactUsLinkComponent
Magnesium sulphate ≥98.0%, AnalaR NORMAPUR® analytical reagent
Magnesium sulphate ≥98.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 25164.265
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium sulphate ≥98.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 25164.265
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Loss on ignition (600°C):
    Max. 2 %
  • Total N (Nitrogen):
    Max. 40 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Ca (Calcium):
    Max. 0.04 %
  • Fe (Iron):
    Max. 10 ppm
  • Mn (Manganese):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :120.37 g/mol
Merck Index:12,05731
Melting Pt:1127 °C
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110