Order Entry
Sweden
ContactUsLinkComponent
 
Magnesium sulphate ≥99.0% (excluding ignited substances), powder USP, Macron Fine Chemicals™
 
undefined
Magnesium sulphate ≥99.0% (excluding ignited substances), powder USP, Macron Fine Chemicals™
 

 

  • GMP Manufactured Product:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤2%
  • Bulk Pharmaceutical Chemical:
    5.0 - 9.2
  • Appearance:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Loss on Drying at 105°C:
    ≤2 %
  • Solubility:
    Passes Test
  • USP - pH (1 in 20):
    5.0 - 9.2
  • USP - Chloride (Cl):
    ≤0.014 %
  • USP - Selenium (Se):
    ≤0.003 %
  • USP - Iron (Fe):
    ≤20 ppm
  • USP - Assay (as MgSO4) (ignited basis):
    99.0 - 100.5 %
  • Thru U.S. No. 100 Sieve:
    ≥85 %