Order Entry
Sweden
ContactUsLinkComponent
Calcium hydroxide, EMPROVE® ESSENTIAL USP, FCC, E526 (max. 0.0005% Al), precipitated, SAFC®
Corrosive
Irritant
Calcium hydroxide, EMPROVE® ESSENTIAL USP, FCC, E526 (max. 0.0005% Al), precipitated, SAFC®
Catalog # 1.02119.9025
CAS Number:  
Corrosive
Irritant
Calcium hydroxide, EMPROVE® ESSENTIAL USP, FCC, E526 (max. 0.0005% Al), precipitated, SAFC®
Catalog # 1.02119.9025
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :74.09 g/mol
Merck Index:13,01675
Melting Pt:550 °C
Density:2.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
UN:3262
ADR:8,III
 :GHS05
 :GHS07
CAS Number:1305-62-0
Formula:Ca(OH)₂
MDL Number:MFCD00010901