Magnesium fluoride ≥99.9% (metals basis), Optical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-40-6
:
Formula:MgF₂
MDL Number:MFCD00011108
:62.3 g/mol
Merck Index:14,5665
Boiling Pt:2260 °C (1013 hPa)
Melting Pt:1261 °C
Density:3.148
EINECS:231-995-1
:GHS07
Storage Temperature:Ambient