Skip to Footer
Avancerad
Avancerad
SDS
Certifikat
Tillverkardata
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcium carbonate
Calcium carbonate 99.95-100.05% ACS chelometric standard
Calcium carbonate 99.95-100.05% ACS chelometric standard
:
Thermo Fisher Scientific
Calcium carbonate 99.95-100.05% ACS chelometric standard
:
Thermo Fisher Scientific
:
471-34-1
:
:
CaCO₃
:
MFCD00010906
:
100.09 g/mol
:
14,1657
:
800 °C (dec.)
:
2.930
:
Ambient
:
207-439-9
msds_ALFA036399
Thermo Scientific Acroseal 25 Years