
Barium carbonate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:513-77-9
:
Formula:BaCO₃
MDL Number:MFCD00003448
:197.34 g/mol
Merck Index:13,00972
Melting Pt:1350 °C
Density:3.89 g/cm³ (20 °C)
EINECS:208-167-3
ADR:6.1,III
:M-Clarity™ quality level = MQ300
UN:1564
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.02 g/l (20 °C)
- Melting Point:1450 °C (decomposition)
- Molar Mass:197.34 g/mol
- Bulk Density:350 kg/m3
- Density:4.43 g/cm3 (20 °C)
- pH:7 - 8 (0.016 g/l, H2O, 16 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials