Magnesium perchlorate
Catalog # GFSC55
Supplier: GFS CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:223.21 g/mol
Merck Index:13,05702
Melting Pt:251 °C
Density:2.21 g/cm³ (20 °C)
EINECS:233-108-3
UN:1475
ADR:5.1,II
:GHS03
:GHS07
CAS Number:10034-81-8
Formula:Cl₂MgO₈
MDL Number:MFCD00011102