

Potassium hydrogen sulphate ≥98%, GPR RECTAPUR®
27012.297
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Assay:Min. 98 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 20 ppm
- PO4 (Phosphate):Max. 20 ppm
- Fe (Iron):Max. 20 ppm
:
:13,07695
:214 °C
:2.32 g/cm³ (20 °C)
:8,II
:136.17 g/mol
:231-594-1
:2509
:GHS05
:GHS07
:7646-93-7
:KHSO₄
:MFCD00011404