Order Entry
Sweden
ContactUsLinkComponent
Kaliumsulfat ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytiskt reagens
Kaliumsulfat ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytiskt reagens
Supplier:  VWR Chemicals
Certificates
undefined
Kaliumsulfat ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytiskt reagens
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • pH (25°C; 5 %):
    5.5 - 7.5
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 100 ppm
  • Total N (Nitrogen):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Mg (Magnesium):
    Max. 20 ppm
  • Na (Sodium):
    Max. 0.02 %
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results