Order Entry
Sweden
ContactUsLinkComponent
 
Corrosive
Irritant
Potassium disulphite ≥96.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 26805.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Potassium disulphite ≥96.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 26805.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 96.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Zn (Zinc):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :222.33 g/mol
Merck Index:13,07729
Melting Pt:190 °C
Density:1.2 g/cm³ (20 °C)
EINECS:240-795-3
 :GHS05
 :GHS07
CAS Number:16731-55-8
Formula:K₂O₅S₂
MDL Number:MFCD00167605