Order Entry
Sweden
ContactUsLinkComponent
 
Potassium hydroxide
Catalog # HONE17851.5000
CAS Number:  
Potassium hydroxide
Catalog # HONE17851.5000
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
 :GHS05
 :GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553