Benzotriazole for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:95-14-7
:
Formula:C₆H₅N₃
MDL Number:MFCD00005699
:119.13 g/mol
Merck Index:13,01109
Boiling Pt:350 °C (1013 hPa)
Melting Pt:99 °C
Density:1.34 g/cm³ (20 °C)
EINECS:202-394-1
UN:1325
ADR:4.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:170 °C (closed cup)
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:19 g/l (20 °C)
- Melting Point:99 °C
- Molar Mass:119.12 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:350 °C (1013 hPa)
- Vapor Pressure:0.053 hPa (20 °C)
- Flash Point:212 °C
- Density:1.36 g/cm3 (20 °C)
- pH:6.0 - 7.0 (100 g/l, H2O, 20 °C) suspension
- Ignition Point:400 °C