Benzo[e]pyrene
Catalog # EHERC20645000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:252.32 g/mol
Merck Index:12,01135
Boiling Pt:492 °C (1013 hPa)
Melting Pt:178…179 °C
EINECS:205-892-7
UN:3077
ADR:9,III
:GHS08
:GHS09
CAS Number:192-97-2
Formula:C₂₀H₁₂