Biphenyl-2-ol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90-43-7
:
Formula:C₁₂H₁₀O
MDL Number:MFCD00002208
:170.21 g/mol
Merck Index:14,07304
Boiling Pt:286°C
Melting Pt:56…58 °C
Density:1,213 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Ambient
EINECS:201-993-5
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:57.0 to 60.0 deg-C
- Solubility in Methanol:almost transparency