Order Entry
Sweden
ContactUsLinkComponent
 
3-Phenylaniline ≥99.0% (by GC, titration analysis)
 :  TCI
 
undefined
3-Phenylaniline ≥99.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to lump
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %