Order Entry
Sweden
ContactUsLinkComponent
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
Catalog # EHERC20011800
Supplier:  EHRENSTORFER
CAS Number:  
2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)
Catalog # EHERC20011800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :326.44 g/mol
UN:2315
ADR:9,II
CAS Number:31508-00-6
Formula:C₁₂H₅Cl₅