4,4'-Dimethylbenzil ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3457-48-5
:
Formula:C₁₆H₁₄O₂
MDL Number:MFCD00008554
:238.29 g/mol
Boiling Pt:209…211 °C (7 mmHg)
Melting Pt:102…104 °C
Storage Temperature:Omgivande
EINECS:222-388-2