Order Entry
Sweden
ContactUsLinkComponent
4-Methylbenzyl alcohol
4-Methylbenzyl alcohol
Catalog # ROTH5263.1
Supplier:  Roth Carl
CAS Number:  
4-Methylbenzyl alcohol
Catalog # ROTH5263.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :122.17 g/mol
Boiling Pt:217 °C (1013 hPa)
Boiling Pt:217 °C (1013 hPa)
Melting Pt:59 °C
Density:0.978 g/cm³ (72 °C)
EINECS:209-639-1
CAS Number:589-18-4
Formula:C₈H₁₀O
MDL Number:MFCD00004664