Order Entry
Sweden
Orders LinkContactUsLinkComponent
Thymol blue

Thymol blue

Glass bottle for solids, 25 g
Catalog # MOLE12900344-25G
Marketsource
Supplier:   Molekula
Thymol blue
Catalog # MOLE12900344-25G
Supplier:  Molekula
Marketsource
CAS Number:  76-61-9
Thymol blue

Glass bottle for solids, 25 g

CAS Number:  76-61-9
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:12,09541
  • Melting Pt:221 °C
  • Storage Temperature:Ambient
  • EINECS:200-973-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.