Order Entry
Sweden
ContactUsLinkComponent
2-Aminobenzophenone 98%
Irritant
2-Aminobenzophenone 98%
Certificates
undefined
Irritant
2-Aminobenzophenone 98%

Specification Test Results

  • Appearance:
    Yellow Powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    105 to 108 °C
  • Titration with HClO4:
    ≥97.5 %
  • Titration by coupling:
    ≥97.0 %

Specification Test Results

Related Information

Frequently Bought Together