
3-Hydroxybenzyl alcohol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:620-24-6
:
Formula:C₇H₈O₂
MDL Number:MFCD00004643
Boiling Pt:86 - 87 °C (0,001 -rr)
Melting Pt:67…71 °C
Density:1,161 g/cm³ (25 °C)
:124.14 g/mol
Storage Temperature:Ambient
EINECS:210-633-6
:GHS05
Specification Test Results
- Appearance:White to beige to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:64 to 71 °C
- GC:≥96.0 %