Order Entry
Sweden
ContactUsLinkComponent
3,5-Dihydroxytoluene
3,5-Dihydroxytoluene
Catalog # MOLE12793573-25G
Supplier:  Molekula
CAS Number:  
3,5-Dihydroxytoluene
Catalog # MOLE12793573-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :124.14 g/mol
Merck Index:12,06995
Boiling Pt:290 °C (1013 hPa)
Melting Pt:109 °C
Density:1.2895 g/cm³ (20 °C)
Flash Pt:159 °C
Storage Temperature:Refrigerator
EINECS:207-984-2
CAS Number:504-15-4
Formula:C₇H₈O₂
MDL Number:MFCD00002291