3,5-Dihydroxytoluene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-15-4
:
Formula:C₇H₈O₂
MDL Number:MFCD00002291
:124.14 g/mol
Merck Index:12,06995
Boiling Pt:290 °C (760 mmHg)
Melting Pt:105…110 °C
Density:1,2895 g/cm³ (20 °C)
Flash Pt:159 °C
EINECS:207-984-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Pink to grey to light brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:105 to 110 °C
- GC:≥97.5 %