
Benzylalkohol 97-100.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
:108.14 g/mol
Merck Index:13,01126
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15,3 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:101 °C
EINECS:202-859-9
:GHS07
Storage Temperature:Omgivande
Specification Test Results
- Assay:97 - 100.5 %
- Acidity:Passes test
- Appearance of solution:Passes test
- IR Spectrum:Passes test
- Density (20/20):1.043 - 1.049
- Peroxide value:Max. 5
- Refractive index (20°C):1.538 - 1.541
- Benzaldehyde:Max. 0.15 %
- Evaporation residue:Max. 0.05 %
- Related substances:Max. 0.2 %
- Cl (Chloride):Max. 20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Packaging Information
spec_VWRC27354EE spec_VWRC27354EE spec_VWRC27354UU
Product specification
VWR Chemicals Big Packs and Bulks