Order Entry
Sweden
ContactUsLinkComponent
3-Phenyl-1-propanol 98%
3-Phenyl-1-propanol 98%
Certificates
undefined
3-Phenyl-1-propanol 98%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5250 to 1.5270 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information