2,4-Dimethoxybenzyl alcohol 99%
:7314-44-5
:
:C₉H₁₂O₃
:MFCD00004614
:168.19 g/mol
:177…179 °C (10 mmHg)
:37…40 °C
:>110 °C
:Ambient
:230-775-2
- Appearance:Clear colorless to light yellow Liquid or low melting solid
- Infrared spectrum:Conforms
- Melting point:34 to 38 °C
- GC:≥98.5 %