Order Entry
Sweden
ContactUsLinkComponent
4-Methoxybenzyl alcohol
4-Methoxybenzyl alcohol
Catalog # MOLE44921023-500G
Supplier:  Molekula
CAS Number:  
4-Methoxybenzyl alcohol
Catalog # MOLE44921023-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :138.17 g/mol
Merck Index:13,00608
Boiling Pt:145 °C (1013 hPa)
Melting Pt:20…24 °C
Density:1.1 g/cm³ (20 °C)
EINECS:203-273-6
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:105-13-5
Formula:C₈H₁₀O₂
MDL Number:MFCD00004653