

3-(Trifluoromethyl)thiophenol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:937-00-8
:
Formula:C₇H₅F₃S
MDL Number:MFCD00041142
:178.18 g/mol
Boiling Pt:71…72 °C (20 mmHg)
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient