Order Entry
Sweden
ContactUsLinkComponent
3,5-Difluorobenzyl alcohol
3,5-Difluorobenzyl alcohol
Catalog # MAYBSB01684EA
Supplier:  Thermo Scientific
CAS Number:  
3,5-Difluorobenzyl alcohol
Catalog # MAYBSB01684EA
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :144.12 g/mol
Boiling Pt:97…99 °C (9 mmHg)
Density:1,27 g/cm³ (20 °C)
 :GHS07
CAS Number:79538-20-8
Formula:C₇H₆F₂O
MDL Number:MFCD00010313