Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-p-toluic acid
2-Fluoro-p-toluic acid ≥97%
2-Fluoro-p-toluic acid ≥97%
Irritant
2-Fluoro-p-toluic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2-Fluoro-p-toluic acid ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7697-23-6
:
Formula:
C₈H₇FO₂
MDL Number:
MFCD03092906
:
154.14 g/mol
Melting Pt:
186…189 °C
EINECS:
None
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH26688
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation