
2'-Chloroacetophenone for synthesis, Sigma-Aldrich®
Catalog # 8.41072.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 ml
- Seq:2370
Specifications
Specification Test Results
- Hill Formula:C₈H₇ClO
- Molar Mass:154.59 g/mol
- Storage class:6.1B Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
- WGK:WGK 3 highly water endangering
- R Phrase:R 22-36/37/38
Specification Test Results
About this item
:
:154.6 g/mol
Boiling Pt:227 °C (1013 hPa)
Density:1.189 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:218-397-6
UN:3416
:GHS07
CAS Number:2142-68-9
Formula:C₈H₇ClO
MDL Number:MFCD00000560