
2'-Chloroacetophenone 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2142-68-9
:
Formula:C₈H₇ClO
MDL Number:MFCD00000560
:154.6 g/mol
Boiling Pt:227 °C (1013 hPa)
Density:1.188 g/cm³
Flash Pt:92 °C
Storage Temperature:Ambient
EINECS:218-397-6
ADR:6.1,II
:GHS07
Specification Test Results
- Appearance:Clear light yellow to amber Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5420 to 1.5450 (20°C, 589 nm)
- GC:≥96.0 %