
2-Bromobenzyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:578-51-8
:
Formula:C₇H₆BrCl
MDL Number:MFCD00040863
:205.48 g/mol
Boiling Pt:110…111 °C (15 mmHg)
Density:1.7341 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05